Assay Details
Target Gene Details
Entrez Gene ID: | 64091 |
Gene Name: | popeye domain containing 2 |
Gene Aliases: |
POP2 |
Location: |
Chr.3:119642052-119660590 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POPDC2 | NM_001308333.1 | 5 | 1462 | NP_001295262.1 |
| NM_022135.3 | 4 | 1379 | NP_071418.2 | |
| XM_005247698.3 | 4 | 1520 | XP_005247755.1 | |
| AF204173.1 | 4 | 1368 | AAG23406.1 | |
| AK124602.1 | 1 | 4614 | ||
| AK226022.1 | 6 | 1668 | ||
| AK377174.1 | 7 | 859 | ||
| AK377176.1 | 7 | 1137 | ||
| AK377177.1 | 5 | 1141 | ||
| AK377178.1 | 7 | 1195 | ||
| AK377179.1 | 6 | 1227 | ||
| AK377180.1 | 8 | 1268 | ||
| AK377181.1 | 7 | 1276 | ||
| AK377182.1 | 7 | 1336 | ||
| AK377183.1 | 8 | 1397 | ||
| AK377184.1 | 8 | 1409 | ||
| BC026911.2 | 6 | 1009 | AAH26911.1 | |
| BC044929.1 | 4 | 1379 | AAH44929.1 | |
| BU948168.1 | 1 | 322 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527724 | Chr.3:119634997 - 119642568 on Build GRCh38 | Loss |
|
| nsv1014394 | Chr.3:119641659 - 119693428 on Build GRCh38 | Loss |
|
| nsv829695 | Chr.3:119460043 - 119649149 on Build GRCh38 | Loss |
|
| nsv520805 | Chr.3:119640198 - 119642568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map