Assay Details
Target Gene Details
Entrez Gene ID: | 11069 |
Gene Name: | Rap guanine nucleotide exchange factor 4 |
Gene Aliases: |
CAMP-GEFII, CGEF2, EPAC, EPAC 2, EPAC2, Nbla00496 |
Location: |
Chr.2:172735282-173052893 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPGEF4 | NM_007023.3 | 1 | 212 | NP_008954.2 |
| XM_005246246.2 | 1 | 79 | XP_005246303.1 | |
| XM_006712205.3 | XP_006712268.1 | |||
| XM_011510505.2 | 1 | 79 | XP_011508807.1 | |
| XM_017003196.1 | 1 | 79 | XP_016858685.1 | |
| AB027471.1 | 1 | 82 | BAB72179.1 | |
| AB209681.1 | 1 | 141 | BAD92918.1 | |
| AK313084.1 | 1 | 81 | ||
| BC024004.2 | 1 | 51 | AAH24004.1 | |
| BI546558.1 | 1 | 172 | ||
| DB471672.1 | 1 | 217 | ||
| KJ534924.1 | ||||
| KJ534925.1 | ||||
| KJ534978.1 | ||||
| U78516.1 | 1 | 130 | AAD03422.1 |
Target Gene Details
Entrez Gene ID: | 91149 |
Gene Name: | RAPGEF4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:172723189-172736206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPGEF4-AS1 | NR_026995.1 | |||
| AL157450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583665 | Chr.2:172595912 - 172778359 on Build GRCh38 | Gain |
|
| esv2760591 | Chr.2:172532177 - 172854792 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map