Assay Details
Target Gene Details
Entrez Gene ID: | 53828 |
Gene Name: | FXYD domain containing ion transport regulator 4 |
Gene Aliases: |
CHIF |
Location: |
Chr.10:43371644-43376335 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXYD4 | NM_001184963.1 | NP_001171892.1 | ||
| NM_173160.2 | NP_775183.1 | |||
| AA759244.1 | ||||
| AI829935.1 | ||||
| AY358584.1 | AAQ88947.1 | |||
| BC054876.1 | AAH54876.1 | |||
| BF507928.1 | ||||
| BG432244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521283 | Chr.10:43371080 - 43376550 on Build GRCh38 | Loss |
|
| nsv831842 | Chr.10:43351093 - 43492053 on Build GRCh38 | Loss |
|
| dgv728n100 | Chr.10:43225695 - 43378216 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map