Assay Details
Target Gene Details
Entrez Gene ID: | 349149 |
Gene Name: | gap junction protein gamma 3 |
Gene Aliases: |
CX29, CX30.2, CX31.3, GJE1 |
Location: |
Chr.7:99923269-99929620 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJC3 | NM_181538.2 | 1 | 337 | NP_853516.1 |
| AF503615.1 | 1 | 337 | AAM21145.1 | |
| AY297109.1 | 1 | 337 | AAP51161.1 | |
| BC043381.1 | AAH43381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607930 | Chr.7:99927618 - 100004568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map