Assay Details
Target Gene Details
Entrez Gene ID: | 10597 |
Gene Name: | trafficking protein particle complex 2B |
Gene Aliases: |
MIP-2A, SEDLP, SEDLP1, TRAPPC2P1 |
Location: |
Chr.19:57363511-57365353 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC2B | NR_002166.1 | |||
| AF058918.1 | AAC14421.1 | |||
| AF291676.1 | ||||
| BC008889.2 | ||||
| BC032809.1 | ||||
| BF984818.1 | 1 | 42 | ||
| BU556832.1 |
Target Gene Details
Entrez Gene ID: | 284306 |
Gene Name: | zinc finger protein 547 |
Gene Aliases: |
- |
Location: |
Chr.19:57363435-57379559 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF547 | NM_173631.3 | 1 | 85 | NP_775902.2 |
| AK055662.1 | BAB70979.1 | |||
| AL832545.1 | ||||
| BC042681.1 | AAH42681.2 |
More Information
Set Membership: |
|

Genomic Map