Assay Details
Target Gene Details
Entrez Gene ID: | 64078 |
Gene Name: | solute carrier family 28 member 3 |
Gene Aliases: |
CNT3 |
Location: |
Chr.9:84273123-84368634 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC28A3 | NM_001199633.1 | NP_001186562.1 | ||
| NM_022127.2 | NP_071410.1 | |||
| NR_037638.2 | ||||
| XM_011518905.2 | XP_011517207.1 | |||
| XM_011518906.2 | XP_011517208.1 | |||
| XM_011518907.2 | XP_011517209.1 | |||
| XM_011518908.2 | XP_011517210.1 | |||
| AF305210.1 | AAG22551.1 | |||
| AK292849.1 | ||||
| AK304406.1 | ||||
| AK314039.1 | ||||
| BC093821.1 | AAH93821.1 | |||
| BC093823.1 | AAH93823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048977 | Chr.9:84149197 - 84815576 on Build GRCh38 | Gain |
|
| nsv8542 | Chr.9:84134385 - 84459995 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map