Assay Details
Target Gene Details
Entrez Gene ID: | 3321 |
Gene Name: | immunoglobulin superfamily member 3 |
Gene Aliases: |
EWI-3, LCDD, V8 |
Location: |
Chr.1:116574398-116667755 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF3 | NM_001007237.2 | 11 | 5961 | NP_001007238.1 |
| NM_001542.3 | 11 | 5883 | NP_001533.2 | |
| XM_005270793.2 | 12 | 5993 | XP_005270850.1 | |
| XM_005270794.4 | 10 | 5231 | XP_005270851.1 | |
| XM_006710593.3 | 11 | 5294 | XP_006710656.1 | |
| XM_011541315.1 | 12 | 6045 | XP_011539617.1 | |
| XM_011541316.2 | 6 | 4774 | XP_011539618.1 | |
| AB007935.2 | 10 | 5293 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482103 | Chr.1:116574409 - 116667692 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map