Assay Details
Target Gene Details
Entrez Gene ID: | 4208 |
Gene Name: | myocyte enhancer factor 2C |
Gene Aliases: |
C5DELq14.3, DEL5q14.3 |
Location: |
Chr.5:88718241-88904105 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
More Information
Set Membership: |
|

Genomic Map