Assay Details
Target Gene Details
Entrez Gene ID: | 55163 |
Gene Name: | pyridoxamine 5'-phosphate oxidase |
Gene Aliases: |
HEL-S-302, PDXPO |
Location: |
Chr.17:47941523-47949308 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPO | NM_018129.3 | 7 | 948 | NP_060599.1 |
| XM_005257500.3 | 7 | 1138 | XP_005257557.1 | |
| XM_011524968.2 | 6 | 721 | XP_011523270.1 | |
| XM_017024813.1 | 7 | 736 | XP_016880302.1 | |
| AF468030.1 | AAM76918.1 | |||
| AK001397.1 | 7 | 900 | BAA91668.1 | |
| AK223242.1 | 7 | 879 | BAD96962.1 | |
| AK303536.1 | 6 | 723 | ||
| AK303665.1 | 6 | 771 | ||
| AK303792.1 | 6 | 847 | ||
| BC006525.2 | 7 | 835 | AAH06525.1 | |
| FJ224333.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1000014 | Chr.17:47941862 - 47952091 on Build GRCh38 | Deletion |
|
| nsv518892 | Chr.17:47810585 - 47952066 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map