Assay Details
Target Gene Details
Entrez Gene ID: | 64783 |
Gene Name: | RNA binding motif protein 15 |
Gene Aliases: |
OTT, OTT1, SPEN |
Location: |
Chr.1:110338426-110373005 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM15 | AB209680.1 | BAD92917.1 |
Target Gene Details
Entrez Gene ID: | 9122 |
Gene Name: | solute carrier family 16 member 4 |
Gene Aliases: |
MCT4, MCT5 |
Location: |
Chr.1:110362851-110391970 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A4 | NM_001201546.1 | 8 | 1570 | NP_001188475.1 |
| NM_001201547.1 | 8 | 1581 | NP_001188476.1 | |
| NM_001201548.1 | 7 | 1457 | NP_001188477.1 | |
| NM_001201549.1 | 8 | 1210 | NP_001188478.1 | |
| NM_001319220.1 | 7 | 1077 | NP_001306149.1 | |
| NM_004696.2 | 9 | 1714 | NP_004687.1 | |
| XM_005271317.4 | 10 | 2732 | XP_005271374.1 | |
| XM_006711033.3 | 7 | 2323 | XP_006711096.1 | |
| AK129985.1 | 9 | 1658 | ||
| AK223307.1 | 9 | 1658 | BAD97027.1 | |
| AK290720.1 | 8 | 1514 | ||
| AK295946.1 | ||||
| AK298539.1 | 7 | 1401 | ||
| AK313735.1 | ||||
| AL833619.1 | 8 | 1511 | ||
| BC021664.1 | 8 | 1180 | AAH21664.1 | |
| BX647948.1 | 7 | 1043 | ||
| U59185.1 | 9 | 1646 | AAB72035.1 |
More Information
Set Membership: |
|

Genomic Map