Assay Details
Target Gene Details
Entrez Gene ID: | 8490 |
Gene Name: | regulator of G-protein signaling 5 |
Gene Aliases: |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129 |
Location: |
Chr.1:163142299-163321791 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS5 | NM_001195303.2 | NP_001182232.1 | ||
| NM_001254748.1 | NP_001241677.1 | |||
| NM_001254749.1 | NP_001241678.1 | |||
| NM_003617.3 | NP_003608.1 | |||
| AB008109.1 | BAA22889.1 | |||
| AF030108.1 | AAB84001.1 | |||
| AF159570.1 | AAD40957.1 | |||
| AF493929.1 | AAM12643.1 | |||
| AJ891044.1 | CAI76926.1 | |||
| AK315357.1 | ||||
| AU130764.1 | ||||
| BC030059.1 | AAH30059.1 | |||
| BP292867.1 | ||||
| BU172751.1 | ||||
| BX537427.1 | CAD97669.1 | |||
| DA731601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890704 | Chr.1:163103712 - 163233339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map