Assay Details
Target Gene Details
Entrez Gene ID: | 2720 |
Gene Name: | galactosidase beta 1 |
Gene Aliases: |
EBP, ELNR1, MPS4B |
Location: |
Chr.3:32996608-33097230 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1 | NM_000404.3 | NP_000395.2 | ||
| NM_001079811.2 | NP_001073279.1 | |||
| NM_001135602.2 | NP_001129074.1 | |||
| NM_001317040.1 | NP_001303969.1 | |||
| AK130133.1 | ||||
| AK222763.1 | BAD96483.1 | |||
| AK223091.1 | BAD96811.1 | |||
| AK298985.1 | ||||
| AK300021.1 | ||||
| AK300753.1 | ||||
| AK312988.1 | ||||
| AU134179.1 | ||||
| BC007493.2 | AAH07493.1 | |||
| BT007147.1 | AAP35811.1 | |||
| BX485230.1 | ||||
| DA077250.1 | ||||
| DA261513.1 | ||||
| DB504658.1 | ||||
| DC301575.1 | ||||
| M22590.1 | AAA51822.1 | |||
| M27507.1 | AAA51819.1 | |||
| M27508.1 | AAA35599.1 | |||
| M34423.1 | AAA51823.1 |
Target Gene Details
Entrez Gene ID: | 643853 |
Gene Name: | transmembrane protein with metallophosphoesterase domain |
Gene Aliases: |
- |
Location: |
Chr.3:33090416-33096801 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPPE | NM_001039770.2 | 2 | 678 | NP_001034859.2 |
| NM_001136238.1 | 2 | 163 | NP_001129710.1 | |
| AK126979.1 | 2 | 678 | BAC86774.1 | |
| AK128242.1 | 2 | 163 | BAC87349.1 | |
| BC136872.1 | 1 | 604 | ||
| BC136873.1 | 1 | 604 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv998302 | Chr.3:32595752 - 33112754 on Build GRCh38 | Gain |
|
| nsv590050 | Chr.3:33060889 - 33117420 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map