Assay Details
Target Gene Details
Entrez Gene ID: | 91977 |
Gene Name: | myozenin 3 |
Gene Aliases: |
CS-3, CS3, FRP3 |
Location: |
Chr.5:150660841-150679368 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOZ3 | NM_001122853.2 | NP_001116325.1 | ||
| NM_133371.4 | NP_588612.2 | |||
| AF480443.1 | AAL79336.1 | |||
| AJ277961.1 | 2 | 656 | CAC81912.1 | |
| AJ300586.1 | CAC83076.1 | |||
| AJ300587.1 | CAC83077.1 | |||
| AK313873.1 | ||||
| BC069380.1 | AAH69380.1 | |||
| BC069553.1 | AAH69553.1 | |||
| BC074870.2 | AAH74870.1 | |||
| BC074871.2 | AAH74871.1 | |||
| DB069077.1 | ||||
| DB277496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
|
| nsv599960 | Chr.5:150636553 - 150685982 on Build GRCh38 | Loss |
|
| nsv428129 | Chr.5:150641519 - 150906180 on Build GRCh38 | Loss |
|
| esv33898 | Chr.5:150657871 - 150766463 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map