Assay Details
Target Gene Details
Entrez Gene ID: | 3188 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H2 (H') |
Gene Aliases: |
FTP3, HNRPH', HNRPH2, hnRNPH' |
Location: |
Chr.X:101408133-101414140 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPH2 | NM_001032393.2 | 2 | 1318 | NP_001027565.1 |
| NM_019597.4 | 2 | 1330 | NP_062543.1 | |
| AK294144.1 | 3 | 1097 | ||
| AK298133.1 | 3 | 955 | ||
| BC130343.1 | 1 | 1120 | ||
| BC130345.1 | 1 | 1120 | ||
| U01923.1 | 2 | 1168 |
Target Gene Details
Entrez Gene ID: | 100529097 |
Gene Name: | RPL36A-HNRNPH2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:101390890-101414140 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL36A-HNRNPH2 | NM_001199973.1 | 5 | 1597 | NP_001186902.1 |
| NM_001199974.1 | 4 | 1474 | NP_001186903.1 |
More Information
Set Membership: |
|

Genomic Map