Assay Details
Target Gene Details
Entrez Gene ID: | 2676 |
Gene Name: | GDNF family receptor alpha 3 |
Gene Aliases: |
GDNFR3 |
Location: |
Chr.5:138252380-138274564 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA3 | NM_001496.3 | NP_001487.2 | ||
| AF051767.1 | AAC24355.1 | |||
| AK297693.1 | ||||
| AK314022.1 | ||||
| AY358997.1 | AAQ89356.1 | |||
| AY359037.1 | AAQ89396.1 | |||
| BC037951.1 | AAH37951.1 | |||
| FJ200487.1 |
More Information
Set Membership: |
|

Genomic Map