Assay Details
Target Gene Details
Entrez Gene ID: | 23759 |
Gene Name: | peptidylprolyl isomerase like 2 |
Gene Aliases: |
CYC4, CYP60, Cyp-60, UBOX7, hCyP-60 |
Location: |
Chr.22:21665984-21698276 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL2 | NM_001317996.1 | NP_001304925.1 | ||
| NM_014337.3 | NP_055152.1 | |||
| NM_148175.2 | NP_680480.1 | |||
| NM_148176.2 | NP_680481.1 | |||
| XM_005261448.3 | XP_005261505.1 | |||
| XM_011530041.2 | XP_011528343.1 | |||
| XM_011530042.2 | XP_011528344.1 | |||
| XM_011530043.2 | XP_011528345.1 | |||
| XM_011530044.2 | XP_011528346.1 | |||
| XM_011530046.2 | XP_011528348.1 | |||
| XM_011530047.2 | XP_011528349.1 | |||
| XM_011530049.2 | XP_011528351.1 | |||
| XM_011530050.2 | XP_011528352.1 | |||
| XM_011530051.2 | XP_011528353.1 | |||
| AF320072.1 | AAK07635.1 | |||
| AK127259.1 | ||||
| AK289545.1 | ||||
| AK299206.1 | ||||
| AL702046.1 | ||||
| BC000022.2 | AAH00022.1 | |||
| BC028385.2 | AAH28385.1 | |||
| CR456548.1 | CAG30434.1 | |||
| DA863607.1 | ||||
| U37219.1 | AAC50376.1 |
More Information
Set Membership: |
|

Genomic Map