Assay Details
Target Gene Details
Entrez Gene ID: | 79713 |
Gene Name: | IGF like family receptor 1 |
Gene Aliases: |
TMEM149, U2AF1L4 |
Location: |
Chr.19:35739250-35742450 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFLR1 | NM_024660.2 | NP_078936.1 | ||
| AK026226.1 | BAB15400.1 | |||
| AX775837.1 | CAE11618.1 | |||
| BC031329.1 | 1 | 119 | AAH31329.1 | |
| BC126481.1 | ||||
| BC126483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515536 | Chr.19:35728624 - 35750059 on Build GRCh38 | Loss |
|
| nsv579422 | Chr.19:35665686 - 35774606 on Build GRCh38 | Loss |
|
| nsv579423 | Chr.19:35730150 - 35792945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map