Assay Details
Target Gene Details
Entrez Gene ID: | 9563 |
Gene Name: | hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase |
Gene Aliases: |
CORTRD1, G6PDH, GDH |
Location: |
Chr.1:9234767-9271337 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H6PD | NM_001282587.1 | NP_001269516.1 | ||
| NM_004285.3 | NP_004276.2 | |||
| XM_005263540.4 | XP_005263597.1 | |||
| XM_006711052.3 | XP_006711115.1 | |||
| XM_017002865.1 | XP_016858354.1 | |||
| AJ012590.1 | CAA10071.1 | |||
| BC081559.1 | AAH81559.1 | |||
| CR749282.1 | CAH18137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv16n100 | Chr.1:9237601 - 9351410 on Build GRCh38 | Gain |
|
| nsv832092 | Chr.1:9126509 - 9304543 on Build GRCh38 | Loss |
|
| dgv12e212 | Chr.1:9243684 - 9344908 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map