Assay Details
Target Gene Details
Entrez Gene ID: | 1603 |
Gene Name: | defender against cell death 1 |
Gene Aliases: |
OST2 |
Location: |
Chr.14:22564907-22589237 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAD1 | NM_001344.3 | 3 | 579 | NP_001335.1 |
| AK223129.1 | 3 | 566 | BAD96849.1 | |
| BC007403.2 | 3 | 551 | AAH07403.1 | |
| BC009798.1 | 3 | 553 | AAH09798.1 | |
| BP198316.1 | ||||
| D15057.1 | 3 | 565 | BAA03650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047315 | Chr.14:22514241 - 22579448 on Build GRCh38 | Gain |
|
| esv3892469 | Chr.14:22030921 - 22600790 on Build GRCh38 | Gain+Loss |
|
| nsv832746 | Chr.14:22506563 - 22681586 on Build GRCh38 | Gain |
|
| nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain |
|
| nsv832748 | Chr.14:22515242 - 22630030 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map