Assay Details
Target Gene Details
Entrez Gene ID: | 10617 |
Gene Name: | STAM binding protein |
Gene Aliases: |
AMSH, MICCAP |
Location: |
Chr.2:73828911-73873656 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834260 | Chr.2:73678979 - 73849393 on Build GRCh38 | Gain |
|
| esv2759060 | Chr.2:73489905 - 73851465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map