Assay Details
Target Gene Details
Entrez Gene ID: | 199746 |
Gene Name: | U2 small nuclear RNA auxiliary factor 1 like 4 |
Gene Aliases: |
U2AF1-RS3, U2AF1L3, U2AF1L3V1, U2AF1RS3, U2af26 |
Location: |
Chr.19:35742527-35745442 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| U2AF1L4 | NM_001040425.2 | 4 | 366 | NP_001035515.1 |
| NM_144987.3 | NP_659424.2 | |||
| NR_110171.1 | 5 | 454 | ||
| NR_110172.1 | ||||
| NR_110173.1 | ||||
| AK092844.1 | BAC03988.1 | |||
| AK094213.1 | ||||
| AK303983.1 | ||||
| AY461582.1 | 5 | 428 | ||
| AY569437.3 | 4 | 335 | AAT72770.3 | |
| BC021186.1 | AAH21186.1 | |||
| BM696851.1 | ||||
| CD175846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515536 | Chr.19:35728624 - 35750059 on Build GRCh38 | Loss |
|
| nsv579422 | Chr.19:35665686 - 35774606 on Build GRCh38 | Loss |
|
| nsv579423 | Chr.19:35730150 - 35792945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map