Assay Details
Target Gene Details
Entrez Gene ID: | 112479 |
Gene Name: | ERI1 exoribonuclease family member 2 |
Gene Aliases: |
EXOD1, ZGRF5 |
Location: |
Chr.16:20780193-20806473 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERI2 | NM_001142725.1 | NP_001136197.1 | ||
| NM_080663.2 | NP_542394.2 | |||
| AB040937.1 | 1 | 884 | BAA96028.1 | |
| AK127391.1 | BAC86956.1 | |||
| AK292594.1 | ||||
| AK304347.1 | ||||
| BC010503.1 | AAH10503.1 | |||
| BX437401.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050009 | Chr.16:20773077 - 20862801 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
| nsv833165 | Chr.16:20718218 - 20886396 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map