Assay Details
Target Gene Details
Entrez Gene ID: | 118491 |
Gene Name: | cilia and flagella associated protein 70 |
Gene Aliases: |
TTC18 |
Location: |
Chr.10:73253758-73361064 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP70 | NM_145170.3 | 15 | 1765 | NP_660153.3 |
| XM_005269489.2 | 14 | 1635 | XP_005269546.1 | |
| XM_006717604.2 | 16 | 3326 | XP_006717667.1 | |
| XM_006717605.2 | 15 | 1728 | XP_006717668.1 | |
| XM_006717609.2 | 13 | 1542 | XP_006717672.1 | |
| XM_006717610.2 | 14 | 3014 | XP_006717673.1 | |
| XM_006717611.3 | 14 | 1666 | XP_006717674.1 | |
| XM_011539212.2 | 11 | 1297 | XP_011537514.1 | |
| XM_017015620.1 | 16 | 3172 | XP_016871109.1 | |
| XM_017015621.1 | 16 | 3169 | XP_016871110.1 | |
| XM_017015622.1 | 16 | 3169 | XP_016871111.1 | |
| XM_017015623.1 | 16 | 3172 | XP_016871112.1 | |
| XM_017015624.1 | 15 | 3091 | XP_016871113.1 | |
| XM_017015625.1 | 16 | 3172 | XP_016871114.1 | |
| XM_017015626.1 | 16 | 3172 | XP_016871115.1 | |
| XM_017015627.1 | 15 | 2962 | XP_016871116.1 | |
| XM_017015628.1 | 14 | 1544 | XP_016871117.1 | |
| XM_017015629.1 | 16 | 3172 | XP_016871118.1 | |
| XM_017015630.1 | 15 | 3246 | XP_016871119.1 | |
| XM_017015631.1 | 11 | 1103 | XP_016871120.1 | |
| XM_017015632.1 | 14 | 3141 | XP_016871121.1 | |
| XM_017015633.1 | 14 | 1611 | XP_016871122.1 | |
| AK308746.1 | 14 | 1653 | ||
| AL833537.1 | 14 | 1571 | CAH10391.1 | |
| BC032856.2 | 15 | 1760 | AAH32856.1 |
More Information
Set Membership: |
|

Genomic Map