Assay Details
Target Gene Details
Entrez Gene ID: | 1495 |
Gene Name: | catenin alpha 1 |
Gene Aliases: |
CAP102, MDPT2 |
Location: |
Chr.5:138753386-138935034 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Gene Details
Entrez Gene ID: | 26045 |
Gene Name: | leucine rich repeat transmembrane neuronal 2 |
Gene Aliases: |
- |
Location: |
Chr.5:138869390-138875368 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRTM2 | NM_015564.2 | 1 | 124 | NP_056379.1 |
| AB007876.1 | 1 | 76 | ||
| AK291064.1 | 1 | 95 | ||
| AK297332.1 | 1 | 98 | ||
| AK309790.1 | 1 | 60 | ||
| DA400927.1 | 1 | 124 |
More Information
Set Membership: |
|

Genomic Map