Assay Details
Target Gene Details
Entrez Gene ID: | 8412 |
Gene Name: | breast cancer anti-estrogen resistance 3 |
Gene Aliases: |
NSP2, SH2D3B |
Location: |
Chr.1:93561741-93847150 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAR3 | NM_001261408.1 | NP_001248337.1 | ||
| NM_001261409.1 | NP_001248338.1 | |||
| NM_001261410.1 | NP_001248339.1 | |||
| NM_001308251.1 | NP_001295180.1 | |||
| NM_003567.3 | NP_003558.1 | |||
| XM_011542251.2 | XP_011540553.1 | |||
| XM_017002480.1 | XP_016857969.1 | |||
| AF124250.1 | AAD28245.1 | |||
| AK027785.1 | ||||
| AK055442.1 | ||||
| AK301570.1 | ||||
| AL833121.1 | ||||
| AY358996.1 | AAQ89355.1 | |||
| BC039895.1 | AAH39895.1 | |||
| U92715.1 | AAC39777.1 |
Target Gene Details
Entrez Gene ID: | 54874 |
Gene Name: | formin binding protein 1 like |
Gene Aliases: |
C1orf39, TOCA1 |
Location: |
Chr.1:93448067-93569671 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNBP1L | XM_011541625.2 | XP_011539927.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011853 | Chr.1:93500893 - 93573448 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map