Assay Details
Target Gene Details
Entrez Gene ID: | 51377 |
Gene Name: | ubiquitin C-terminal hydrolase L5 |
Gene Aliases: |
CGI-70, INO80R, UCH-L5, UCH37 |
Location: |
Chr.1:193012366-193060140 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv498n100 | Chr.1:192992568 - 193114915 on Build GRCh38 | Loss |
|
| nsv548658 | Chr.1:192394185 - 193552205 on Build GRCh38 | Loss |
|
| nsv498699 | Chr.1:193001306 - 193024494 on Build GRCh38 | Loss |
|
| nsv3966 | Chr.1:193007327 - 193035756 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map