Assay Details
Target Gene Details
Entrez Gene ID: | 60437 |
Gene Name: | cadherin 26 |
Gene Aliases: |
VR20 |
Location: |
Chr.20:59957764-60035629 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH26 | NM_177980.2 | 1 | 110 | NP_817089.1 |
| XM_011528969.2 | 1 | 110 | XP_011527271.1 | |
| XM_017027994.1 | 1 | 761 | XP_016883483.1 | |
| XM_017027995.1 | 1 | 762 | XP_016883484.1 | |
| XM_017027996.1 | 1 | 762 | XP_016883485.1 | |
| XM_017027997.1 | 1 | 762 | XP_016883486.1 | |
| XM_017027998.1 | 1 | 759 | XP_016883487.1 | |
| XM_017027999.1 | 1 | 759 | XP_016883488.1 | |
| XM_017028000.1 | 1 | 110 | XP_016883489.1 | |
| XM_017028001.1 | 1 | 759 | XP_016883490.1 | |
| XM_017028002.1 | 1 | 760 | XP_016883491.1 | |
| AK055202.1 | 1 | 99 | ||
| DB218314.1 | 1 | 110 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1009769 | Chr.20:59957379 - 59963538 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map