Assay Details
Target Gene Details
Entrez Gene ID: | 51700 |
Gene Name: | cytochrome b5 reductase 2 |
Gene Aliases: |
B5R.2 |
Location: |
Chr.11:7665095-7678568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5R2 | NM_001302826.1 | NP_001289755.1 | ||
| NM_001302827.1 | NP_001289756.1 | |||
| NM_016229.4 | NP_057313.2 | |||
| NR_126508.1 | ||||
| XM_005252975.4 | XP_005253032.1 | |||
| XM_006718251.2 | XP_006718314.1 | |||
| XM_011520184.2 | XP_011518486.1 | |||
| XM_011520185.2 | XP_011518487.1 | |||
| XM_017017920.1 | XP_016873409.1 | |||
| XM_017017921.1 | XP_016873410.1 | |||
| XM_017017922.1 | XP_016873411.1 | |||
| AB209000.1 | 2 | 4665 | BAD92237.1 | |
| AF169802.1 | AAF04811.1 | |||
| AK022556.1 | 1 | 872 | ||
| AK290102.1 | ||||
| AK308543.1 | 6 | 1132 | ||
| AL133582.1 | CAB63726.1 | |||
| AY665398.1 | AAT75296.1 | |||
| BF061336.1 | ||||
| BG495762.1 | ||||
| BG723242.1 | ||||
| BM693762.1 | ||||
| BQ431043.1 | ||||
| CB149369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv184e214 | Chr.11:7666840 - 7787334 on Build GRCh38 | Gain |
|
| esv3625293 | Chr.11:7515685 - 7685956 on Build GRCh38 | Gain |
|
| esv3891925 | Chr.11:7596802 - 7700323 on Build GRCh38 | Gain |
|
| esv3625296 | Chr.11:7635550 - 7688660 on Build GRCh38 | Gain |
|
| nsv1035575 | Chr.11:7648656 - 7744807 on Build GRCh38 | Gain |
|
| dgv58n21 | Chr.11:7652157 - 7688219 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map