Assay Details
Target Gene Details
Entrez Gene ID: | 126328 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A11 |
Gene Aliases: |
B14.7, CI-B14.7 |
Location: |
Chr.19:5891276-5904013 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA11 | NM_001193375.1 | NP_001180304.1 | ||
| NM_175614.4 | NP_783313.1 | |||
| NR_034166.2 | ||||
| AJ539081.1 | CAD62165.1 | |||
| AK127692.1 | BAC87088.1 | |||
| AL546175.3 | ||||
| AY726561.1 | ||||
| BC069045.1 | AAH69045.1 | |||
| BI160094.1 | ||||
| BP432135.1 | ||||
| BQ054257.1 | ||||
| BQ671956.1 | ||||
| BQ955016.1 |
More Information
Set Membership: |
|

Genomic Map