Assay Details
Target Gene Details
Entrez Gene ID: | 63826 |
Gene Name: | serine racemase |
Gene Aliases: |
ILV1, ISO1 |
Location: |
Chr.17:2303371-2325267 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRR | NM_001304803.1 | NP_001291732.1 | ||
| NM_021947.2 | NP_068766.1 | |||
| XM_006721565.3 | XP_006721628.1 | |||
| XM_006721566.3 | XP_006721629.1 | |||
| XM_011523974.2 | XP_011522276.1 | |||
| AF169974.1 | AAG27081.1 | |||
| AK023169.1 | BAB14442.1 | |||
| AK223120.1 | BAD96840.1 | |||
| AL834378.1 | CAD39041.1 | |||
| AY034081.1 | AAK58495.1 | |||
| AY743705.1 | AAW66460.1 | |||
| AY743706.1 | AAW66461.1 | |||
| AY743707.1 | AAW66462.1 | |||
| AY743708.1 | AAW66463.1 | |||
| BC074728.2 | AAH74728.1 | |||
| CR457300.1 | CAG33581.1 | |||
| DA297969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574224 | Chr.17:2269769 - 2423186 on Build GRCh38 | Gain |
|
| nsv469765 | Chr.17:2213155 - 2400446 on Build GRCh38 | Loss |
|
| nsv833336 | Chr.17:2176922 - 2400456 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map