Assay Details
Target Gene Details
Entrez Gene ID: | 84658 |
Gene Name: | adhesion G protein-coupled receptor E3 |
Gene Aliases: |
EMR3 |
Location: |
Chr.19:14600175-14674918 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE3 | AL832663.1 |
Target Gene Details
Entrez Gene ID: | 388512 |
Gene Name: | C-type lectin domain family 17 member A |
Gene Aliases: |
- |
Location: |
Chr.19:14580777-14612035 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC17A | NM_001204118.1 | 14 | 1519 | NP_001191047.1 |
| NM_207390.3 | 13 | 1409 | NP_997273.3 | |
| NR_109784.1 | 14 | 1582 | ||
| NR_109785.1 | 13 | 1367 | ||
| XM_017026785.1 | 15 | 2063 | XP_016882274.1 | |
| XM_017026786.1 | 15 | 1927 | XP_016882275.1 | |
| XM_017026787.1 | 16 | 2282 | XP_016882276.1 | |
| XM_017026788.1 | 15 | 2039 | XP_016882277.1 | |
| XM_017026789.1 | 11 | 1741 | XP_016882278.1 | |
| XM_017026790.1 | 14 | 2015 | XP_016882279.1 | |
| XM_017026791.1 | 13 | 1864 | XP_016882280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map