Assay Details
Target Gene Details
Entrez Gene ID: | 92126 |
Gene Name: | dermatan sulfate epimerase-like |
Gene Aliases: |
C18orf4 |
Location: |
Chr.18:67506582-67516730 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSEL | NM_032160.2 | 2 | 3174 | NP_115536.1 |
| AF480435.1 | 2 | 3427 | AAN32895.1 | |
| AK021539.1 | 1 | 1342 | ||
| AK021849.1 | BAB13912.1 | |||
| AK023067.1 | 1 | 994 | ||
| BC062557.1 | 1 | 2258 | AAH62557.1 | |
| BC117325.1 | 1 | 1988 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577302 | Chr.18:67288152 - 67820837 on Build GRCh38 | Loss |
|
| nsv577312 | Chr.18:67481097 - 67552997 on Build GRCh38 | Loss |
|
| nsv1057236 | Chr.18:67395980 - 67782432 on Build GRCh38 | Loss |
|
| esv3642903 | Chr.18:67457986 - 67599133 on Build GRCh38 | Gain |
|
| dgv3383n100 | Chr.18:67133808 - 67550267 on Build GRCh38 | Gain |
|
| nsv577157 | Chr.18:65715200 - 68002467 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map