Assay Details
Target Gene Details
Entrez Gene ID: | 11180 |
Gene Name: | WD repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.3:49007062-49015953 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR6 | NM_001320546.1 | 6 | 3560 | NP_001307475.1 |
| NM_001320547.1 | 6 | 2931 | NP_001307476.1 | |
| NM_018031.4 | 6 | 3346 | NP_060501.3 | |
| AF099100.1 | 6 | 3015 | AAF80244.1 | |
| AK001080.1 | BAA91497.1 | |||
| AK022719.1 | 5 | 1251 | BAB14201.1 | |
| AK092079.1 | 6 | 671 | ||
| AK225653.1 | 5 | 3124 | ||
| AK295145.1 | 6 | 3346 | ||
| AK297090.1 | 6 | 1356 | ||
| AK304117.1 | 6 | 2931 | ||
| AK309913.1 | 6 | 3226 | ||
| AL133589.1 | 5 | 1126 | CAB63730.1 | |
| AL833910.1 | 5 | 2192 | CAD38766.1 | |
| BC002826.2 | 8 | 1027 | AAH02826.1 | |
| BC078176.1 | 5 | 1065 | AAH78176.1 | |
| BC101707.1 | 6 | 3005 | AAI01708.1 | |
| BC113467.1 | 6 | 3005 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954488 | Chr.3:49002068 - 49019667 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map