Assay Details
Target Gene Details
Entrez Gene ID: | 1106 |
Gene Name: | chromodomain helicase DNA binding protein 2 |
Gene Aliases: |
EEOC |
Location: |
Chr.15:92900321-93028007 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD2 | NM_001042572.2 | 3 | 824 | NP_001036037.1 |
| NM_001271.3 | 3 | 824 | NP_001262.3 | |
| AF006514.1 | 3 | 956 | AAB87382.1 | |
| AK292850.1 | 3 | 821 | ||
| AK295909.1 | 2 | 723 | ||
| BC007347.2 | 3 | 811 | AAH07347.1 | |
| BC020810.1 | 3 | 801 | AAH20810.1 | |
| BT007050.1 | 2 | 249 | AAP35699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2660n100 | Chr.15:92869548 - 92932648 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map