Assay Details
Target Gene Details
Entrez Gene ID: | 84678 |
Gene Name: | lysine demethylase 2B |
Gene Aliases: |
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2 |
Location: |
Chr.12:121402289-121582268 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2B | NM_001005366.1 | 23 | 4760 | NP_001005366.1 |
| NM_032590.4 | 23 | 4842 | NP_115979.3 | |
| XM_005253955.3 | 23 | 5052 | XP_005254012.1 | |
| XM_005253956.3 | 23 | 4649 | XP_005254013.1 | |
| XM_005253961.4 | 12 | 3137 | XP_005254018.1 | |
| XM_011538867.2 | XP_011537169.1 | |||
| XM_011538868.2 | 23 | 6100 | XP_011537170.1 | |
| XM_011538869.2 | 24 | 5924 | XP_011537171.1 | |
| XM_011538875.2 | 12 | 3146 | XP_011537177.1 | |
| AB031230.1 | 15 | 3323 | BAA97672.1 | |
| AK027440.1 | 14 | 3118 | BAB55112.1 | |
| AK027692.1 | 10 | 2747 | BAB55301.1 | |
| AK127328.1 | 23 | 4760 | ||
| AL133572.1 | 15 | 6637 | CAB63721.2 | |
| BC008735.2 | 9 | 2612 | AAH08735.2 | |
| CA430876.1 | 1 | 481 |
More Information
Set Membership: |
|

Genomic Map