Assay Details
Target Gene Details
Entrez Gene ID: | 133015 |
Gene Name: | PARK2 coregulated like |
Gene Aliases: |
C4orf28 |
Location: |
Chr.4:20696282-20757766 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829876 | Chr.4:20637113 - 20833998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map