Assay Details
Target Gene Details
Entrez Gene ID: | 84769 |
Gene Name: | MPV17 mitochondrial inner membrane protein like 2 |
Gene Aliases: |
FKSG24 |
Location: |
Chr.19:18193230-18196742 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPV17L2 | NM_032683.2 | NP_116072.2 | ||
| AF320622.1 | AAL30173.1 | |||
| AF416712.1 | AAL16806.1 | |||
| AK094091.1 | ||||
| BC005064.1 | AAH05064.1 | |||
| BC093008.1 | AAH93008.1 | |||
| BG740168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv576n67 | Chr.19:18178788 - 18200792 on Build GRCh38 | Gain |
|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv833775 | Chr.19:18102951 - 18279264 on Build GRCh38 | Loss |
|
| esv3643833 | Chr.19:18192120 - 18200529 on Build GRCh38 | Loss |
|
| nsv819691 | Chr.19:18183343 - 18201465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map