Assay Details
Target Gene Details
Entrez Gene ID: | 2582 |
Gene Name: | UDP-galactose-4-epimerase |
Gene Aliases: |
SDR1E1 |
Location: |
Chr.1:23795599-23800804 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALE | NM_000403.3 | NP_000394.2 | ||
| NM_001008216.1 | NP_001008217.1 | |||
| NM_001127621.1 | NP_001121093.1 | |||
| AK057302.1 | ||||
| AK314397.1 | ||||
| AL530475.3 | ||||
| BC001273.1 | AAH01273.1 | |||
| BC050685.2 | AAH50685.2 | |||
| BQ437612.1 | ||||
| DQ233668.1 | ABB04110.1 | |||
| L41668.1 | AAB86498.1 |
More Information
Set Membership: |
|

Genomic Map