Assay Details
Target Gene Details
Entrez Gene ID: | 5737 |
Gene Name: | prostaglandin F receptor |
Gene Aliases: |
FP |
Location: |
Chr.1:78490974-78540701 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGFR | NM_000959.3 | 1 | 93 | NP_000950.1 |
| NM_001039585.1 | 1 | 93 | NP_001034674.1 | |
| XM_017001873.1 | 1 | 162 | XP_016857362.1 | |
| AF004021.1 | AAB63152.1 | |||
| AK292845.1 | 1 | 77 | ||
| AK312816.1 | 1 | 77 | ||
| AY485530.1 | 1 | 64 | AAR84381.1 | |
| BC112965.2 | AAI12966.1 | |||
| CD013876.1 | 1 | 97 | ||
| L24470.1 | 1 | 64 | AAA17684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1566 | Chr.1:78486860 - 78495143 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map