Assay Details
Target Gene Details
Entrez Gene ID: | 8891 |
Gene Name: | eukaryotic translation initiation factor 2B subunit gamma |
Gene Aliases: |
EIF-2B, EIF2Bgamma |
Location: |
Chr.1:44850522-44986722 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2B3 | NM_001166588.2 | NP_001160060.1 | ||
| NM_001261418.1 | NP_001248347.1 | |||
| NM_020365.4 | NP_065098.1 | |||
| XM_017002745.1 | XP_016858234.1 | |||
| XM_017002746.1 | XP_016858235.1 | |||
| XM_017002747.1 | XP_016858236.1 | |||
| AF257077.1 | AAF91351.1 | |||
| AK022399.1 | BAB14027.1 | |||
| AK024006.1 | BAB14770.1 | |||
| AK314668.1 | ||||
| AL834288.1 | CAD38962.1 | |||
| AW972575.1 | ||||
| BC018728.1 | AAH18728.1 | |||
| CR457285.1 | CAG33566.1 | |||
| CX781577.1 |
More Information
Set Membership: |
|

Genomic Map