Assay Details
Target Gene Details
Entrez Gene ID: | 5306 |
Gene Name: | phosphatidylinositol transfer protein alpha |
Gene Aliases: |
HEL-S-36, PI-TPalpha, PITPN, VIB1A |
Location: |
Chr.17:1517989-1562816 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNA | NM_006224.3 | 12 | 2538 | NP_006215.1 |
| XM_011523939.1 | 11 | 2450 | XP_011522241.1 | |
| XM_011523940.1 | 12 | 2960 | XP_011522242.1 | |
| AL117644.1 | 1 | 305 | ||
| BC045108.1 | 12 | 2513 | AAH45108.1 | |
| BC068538.1 | 12 | 2421 | AAH68538.1 | |
| BC082976.1 | 12 | 2453 | AAH82976.1 | |
| EU794647.1 | 12 | 2513 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142059189] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map