Assay Details
Target Gene Details
Entrez Gene ID: | 1665 |
Gene Name: | DEAH-box helicase 15 |
Gene Aliases: |
DBP1, DDX15, HRH2, PRP43, PRPF43, PrPp43p |
Location: |
Chr.4:24527464-24584603 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX15 | NM_001358.2 | 14 | 2937 | NP_001349.2 |
| AA771922.1 | 1 | 83 | ||
| AB001636.1 | 14 | 2930 | BAA23987.1 | |
| AK054614.1 | 2 | 1782 | ||
| AK055898.1 | 3 | 2005 | ||
| AK056726.1 | 4 | 1945 | ||
| BC035974.1 | 14 | 2924 | AAH35974.1 |
More Information
Set Membership: |
|

Genomic Map