Assay Details
Target Gene Details
Entrez Gene ID: | 389840 |
Gene Name: | mitogen-activated protein kinase kinase kinase 15 |
Gene Aliases: |
ASK3, bA723P2.3 |
Location: |
Chr.X:19360058-19515459 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K15 | NM_001001671.3 | 27 | 3742 | NP_001001671.3 |
| XM_011545507.2 | 28 | 3969 | XP_011543809.2 | |
| XM_011545508.2 | 28 | 3882 | XP_011543810.2 | |
| XM_011545510.2 | 21 | 4482 | XP_011543812.1 | |
| XM_011545511.2 | 18 | 2993 | XP_011543813.1 | |
| AK122632.1 | 6 | 1160 | ||
| AK131412.1 | 24 | 3070 | BAD18559.1 | |
| AK131477.1 | 28 | 3802 | BAD18622.1 |
Target Gene Details
Entrez Gene ID: | 5160 |
Gene Name: | pyruvate dehydrogenase (lipoamide) alpha 1 |
Gene Aliases: |
PDHA, PDHAD, PDHCE1A, PHE1A |
Location: |
Chr.X:19343893-19361707 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDHA1 | NM_000284.3 | 11 | 3196 | NP_000275.1 |
| NM_001173454.1 | 12 | 3310 | NP_001166925.1 | |
| NM_001173455.1 | 11 | 3217 | NP_001166926.1 | |
| NM_001173456.1 | 10 | 3103 | NP_001166927.1 | |
| AW204705.1 | 1 | 169 | ||
| R49470.1 | 1 | 195 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573627 | Chr.X:19351701 - 19362562 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map