Assay Details
Target Gene Details
Entrez Gene ID: | 2793 |
Gene Name: | G protein subunit gamma transducin 2 |
Gene Aliases: |
G-GAMMA-8, G-GAMMA-C, GNG8, GNG9, GNGT8 |
Location: |
Chr.17:49206234-49210574 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNGT2 | NM_001198754.1 | NP_001185683.1 | ||
| NM_001198755.1 | NP_001185684.1 | |||
| NM_001198756.1 | NP_001185685.1 | |||
| NM_031498.2 | 1 | 120 | NP_113686.1 | |
| AK312839.1 | ||||
| BC008663.1 | 1 | 120 | AAH08663.1 | |
| BM723861.1 | ||||
| DA248836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2074 | Chr.17:49208171 - 49233994 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map