Assay Details
Target Gene Details
Entrez Gene ID: | 51279 |
Gene Name: | complement C1r subcomponent like |
Gene Aliases: |
C1RL1, C1RLP, C1r-LP, CLSPa |
Location: |
Chr.12:7093662-7109278 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1RL | NM_001297640.1 | 2 | 314 | NP_001284569.1 |
| NM_001297642.1 | 2 | 314 | NP_001284571.1 | |
| NM_001297643.1 | 2 | 314 | NP_001284572.1 | |
| NM_016546.3 | 2 | 314 | NP_057630.2 | |
| XM_017019397.1 | 2 | 361 | XP_016874886.1 | |
| XM_017019398.1 | 2 | 361 | XP_016874887.1 | |
| XM_017019399.1 | 2 | 361 | XP_016874888.1 | |
| XM_017019400.1 | 2 | 361 | XP_016874889.1 | |
| XM_017019401.1 | 2 | 361 | XP_016874890.1 | |
| XM_017019402.1 | 2 | 361 | XP_016874891.1 | |
| XM_017019403.1 | 2 | 361 | XP_016874892.1 | |
| XM_017019404.1 | 2 | 361 | XP_016874893.1 | |
| AF178985.1 | 2 | 233 | AAF44349.1 | |
| AK024084.1 | 2 | 296 | BAB14819.1 | |
| AK222802.1 | 2 | 241 | BAD96522.1 | |
| AK314299.1 | 2 | 274 | ||
| BC062428.1 | 2 | 250 | AAH62428.1 | |
| DB113812.1 | 2 | 309 | ||
| DR004214.1 | 2 | 240 | ||
| HY062376.1 | 2 | 314 |
Target Gene Details
Entrez Gene ID: | 283314 |
Gene Name: | C1RL antisense RNA 1 |
Gene Aliases: |
MATL2963 |
Location: |
Chr.12:7108308-7121851 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1RL-AS1 | NR_026947.1 | |||
| AL833364.1 |
More Information
Set Membership: |
|

Genomic Map