Assay Details
Target Gene Details
Entrez Gene ID: | 2220 |
Gene Name: | ficolin 2 |
Gene Aliases: |
EBP-37, FCNL, P35, ficolin-2 |
Location: |
Chr.9:134864146-134890506 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCN2 | NM_004108.2 | NP_004099.2 | ||
| NM_015837.2 | NP_056652.1 | |||
| XM_006717015.3 | 7 | 1367 | XP_006717078.1 | |
| XM_011518392.2 | 8 | 1483 | XP_011516694.1 | |
| AI052239.1 | 1 | 41 | ||
| AK096112.1 | ||||
| AK290843.1 | 8 | 1079 | ||
| AK308476.1 | ||||
| D49353.1 | BAA08352.1 | |||
| JB054537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv1043726 | Chr.9:134566621 - 135388584 on Build GRCh38 | Gain |
|
| nsv831753 | Chr.9:134885467 - 135039086 on Build GRCh38 | Loss |
|
| nsv951815 | Chr.9:134879855 - 134896854 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map