Assay Details
Target Gene Details
Entrez Gene ID: | 65264 |
Gene Name: | ubiquitin conjugating enzyme E2 Z |
Gene Aliases: |
HOYS7, USE1 |
Location: |
Chr.17:48908369-48929060 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2Z | NM_023079.4 | 3 | 643 | NP_075567.2 |
| AB025426.1 | 2 | 184 | BAB87810.1 | |
| AK023917.1 | 2 | 181 | BAB14724.1 | |
| AK024030.1 | 4 | 498 | BAB14789.1 | |
| AK091644.1 | 1 | 1773 | ||
| AK122770.1 | 3 | 541 | ||
| AK130303.1 | 4 | 501 | ||
| AK296865.1 | 4 | 534 | ||
| BC015169.1 | 2 | 170 | AAH15169.2 | |
| BC015890.1 | 3 | 511 | AAH15890.1 | |
| CR457322.1 | 2 | 184 | CAG33603.1 | |
| EF623992.1 | 3 | 508 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain |
|
| nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map