Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf60 |
Target Gene Details
Entrez Gene ID: | 2984 |
Gene Name: | guanylate cyclase 2C |
Gene Aliases: |
DIAR6, GUC2C, MECIL, MUCIL, STAR |
Location: |
Chr.12:14612632-14696625 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY2C | NM_004963.3 | NP_004954.2 | ||
| XM_011520631.2 | XP_011518933.1 | |||
| BC136544.1 | ||||
| BC136545.1 | ||||
| M73489.1 | AAA36655.1 | |||
| S57551.1 | AAB19934.2 |
Target Gene Details
Entrez Gene ID: | 101928290 |
Gene Name: | PLBD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:14567732-14619755 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLBD1-AS1 | NR_120465.1 | |||
| AI922058.1 | ||||
| CB048614.1 | ||||
| DB284711.1 |
More Information
Set Membership: |
|

Genomic Map