Assay Details
Target Gene Details
Entrez Gene ID: | 10884 |
Gene Name: | mitochondrial ribosomal protein S30 |
Gene Aliases: |
MRP-S30, PAP, PDCD9, S30mt |
Location: |
Chr.5:44808925-44815514 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS30 | NM_016640.3 | 1 | 547 | NP_057724.2 |
| AF146192.2 | 1 | 547 | AAF65227.1 | |
| AF217523.1 | 1 | 246 | AAF67634.1 | |
| AK074777.1 | 1 | 515 | BAC11202.1 | |
| AK222704.1 | 1 | 525 | BAD96424.1 | |
| AK222894.1 | 1 | 517 | BAD96614.1 | |
| AL136706.1 | 1 | 520 | CAB66641.1 | |
| AL355715.1 | 1 | 547 | CAB90810.1 | |
| AL355716.1 | 1 | 240 | CAB90811.1 | |
| BC002460.2 | 1 | 498 | AAH02460.2 | |
| BC007735.2 | 1 | 524 | AAH07735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830279 | Chr.5:44688073 - 44860713 on Build GRCh38 | Gain |
|
| nsv597933 | Chr.5:44740887 - 44810638 on Build GRCh38 | Loss |
|
| nsv597934 | Chr.5:44803647 - 44834131 on Build GRCh38 | Loss |
|
| nsv1027897 | Chr.5:44695176 - 45061521 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map