Assay Details
Target Gene Details
Entrez Gene ID: | 140766 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 14 |
Gene Aliases: |
- |
Location: |
Chr.10:70672518-70762441 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS14 | NM_080722.4 | NP_542453.2 | ||
| NM_139155.3 | NP_631894.2 | |||
| XM_011539300.2 | XP_011537602.1 | |||
| XM_011539303.2 | XP_011537605.1 | |||
| XM_011539305.2 | XP_011537607.1 | |||
| XM_011539306.2 | XP_011537608.1 | |||
| XM_011539307.2 | XP_011537609.1 | |||
| XM_011539308.2 | XP_011537610.1 | |||
| AF358666.1 | AAL40229.1 | |||
| AF366351.1 | AAL79814.1 | |||
| AJ345098.1 | CAC87943.1 | |||
| CN398634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| dgv160n67 | Chr.10:70665822 - 70698742 on Build GRCh38 | Gain |
|
| nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss |
|
| nsv831909 | Chr.10:70604167 - 70772302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map